Gene Transcripts
- Gene Symbol
- 4933406B15Rik
- Entrez Gene ID
- 71064
- Gene Name
- RIKEN cDNA 4933406B15 gene
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.410795
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK016679.1 |
- |
2 |
756 |
|
View Full Product Details