Gene Transcripts
- Gene Symbol
- Gm5108
- Entrez Gene ID
- 330097
- Gene Name
- predicted gene 5108
- Gene Aliases
- EG330097
- Chromosome Location
- Chr. 5: 67941669 - 67977070 on Build GRCm38
- UniGene ID
- Mm.78496
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK076825.1 |
- |
1 |
150 |
|
BC120772.1 |
- |
1 |
78 |
|
BC120798.1 |
- |
1 |
78 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AV264477.2 |
- |
1 |
150 |
|
Chr. 5 - 67941669 - 67977070 [+] |
GenBankmRNA |
BB615379.1 |
- |
1 |
150 |
|
Chr. 5 - 67941669 - 67977072 [+] |
View Full Product Details