Gene Transcripts
- Gene Symbol
- 8030442B05Rik
- Entrez Gene ID
- 77543
- Gene Name
- RIKEN cDNA 8030442B05 gene
- Gene Aliases
- Chromosome Location
- Chr. 2: 11338366 - 11398500 on Build GRCm38
- UniGene ID
- Mm.490670
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_040615.1 |
|
3 |
810 |
|
GenBank mRNA |
AK020198.1 |
- |
3 |
811 |
|
View Full Product Details