Gene Transcripts
- Gene Symbol
- Gm16136
- Entrez Gene ID
- 100503499
- Gene Name
- predicted gene 16136
- Gene Aliases
- OTTMUSG00000030878
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.391911
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK005410.1 |
- |
1 |
851 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY705616.1 |
- |
1 |
851 |
|
Chr. 15 - 37232143 - 37233141 [-] |
View Full Product Details