Gene Transcripts
- Gene Symbol
- 9530052C20Rik
- Entrez Gene ID
- 320229
- Gene Name
- RIKEN cDNA 9530052C20 gene
- Gene Aliases
- Chromosome Location
- Chr. 3: 142878399 - 142898015 on Build GRCm38
- UniGene ID
- Mm.187582
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK035466.1 |
- |
2 |
1322 |
|
AK085299.1 |
- |
2 |
1601 |
|
View Full Product Details