Gene Transcripts
- Gene Symbol
- Gm16740
- Entrez Gene ID
- 100504029
- Gene Name
- predicted gene, 16740
- Gene Aliases
- Chromosome Location
- Chr. 3: 95217457 - 95226658 on Build GRCm38
- UniGene ID
- Mm.478537
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_108026.1 |
|
1, 2 |
611 |
|
GenBank mRNA |
AK020406.1 |
- |
1, 2 |
612 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB366280.1 |
- |
1, 2 |
206 |
|
Chr. 3 - 95217862 - 95219544 [+] |
GenBankmRNA |
BF730312.1 |
- |
1, 2 |
263 |
|
Chr. 3 - 95217805 - 95219549 [+] |
GenBankmRNA |
DV065665.1 |
- |
|
599 |
|
Chr. - - [-] |
GenBankmRNA |
BY721575.1 |
- |
1, 2 |
594 |
|
Chr. 3 - 95217474 - 95219494 [+] |
View Full Product Details