Gene Transcripts
- Gene Symbol
- 3110040K02Rik
- Entrez Gene ID
- 73140
- Gene Name
- RIKEN cDNA 3110040K02 gene
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK014159.1 |
- |
1 |
85 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY713134.1 |
- |
1 |
85 |
|
Chr. 2 - 158411515 - 158412002 [+] |
View Full Product Details