Gene Transcripts
- Gene Symbol
- D230034L24Rik
- Entrez Gene ID
- 106251
- Gene Name
- RIKEN cDNA D230034L24 gene
- Gene Aliases
- AI664004
- Chromosome Location
- Chr. : - on Build
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK084380.1 |
- |
1 |
4033 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB470662.2 |
- |
1 |
447 |
|
Chr. 16 - 72801390 - 72801999 [+] |
View Full Product Details