Gene Transcripts
- Gene Symbol
- Kif24
- Entrez Gene ID
- 109242
- Gene Name
- kinesin family member 24
- Gene Aliases
- 4933425J19Rik, 9430029L23Rik
- Chromosome Location
- Chr. 4: 41390748 - 41464933 on Build GRCm38
- UniGene ID
- Mm.370288
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK020445.1 |
- |
2 |
231 |
|
- Gene Symbol
- A630075K04Rik
- Entrez Gene ID
- 319299
- Gene Name
- RIKEN cDNA A630075K04 gene
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.157648
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK042254.1 |
- |
|
3336 |
|
- Gene Symbol
- AU022855
- Entrez Gene ID
- 107295
- Gene Name
- expressed sequence AU022855
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK080354.1 |
- |
1 |
2595 |
|
- Gene Symbol
- Abhd2
- Entrez Gene ID
- 54608
- Gene Name
- abhydrolase domain containing 2
- Gene Aliases
- 2210009N18Rik, LABH2, Labh-2
- Chromosome Location
- Chr. 7: 79273266 - 79361601 on Build GRCm38
- UniGene ID
- Mm.365490
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK053460.1 |
- |
6 |
879 |
|
- Gene Symbol
- Fmo1
- Entrez Gene ID
- 14261
- Gene Name
- flavin containing monooxygenase 1
- Gene Aliases
- Chromosome Location
- Chr. 1: 162829561 - 162866548 on Build GRCm38
- UniGene ID
- Mm.976
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK042457.1 |
- |
4, 5 |
658 |
|
- Gene Symbol
- Kcnh6
- Entrez Gene ID
- 192775
- Gene Name
- potassium voltage-gated channel, subfamily H (eag-related), member 6
- Gene Aliases
- m-erg2
- Chromosome Location
- Chr. 11: 106007994 - 106035595 on Build GRCm38
- UniGene ID
- Mm.343850
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK038513.1 |
- |
7 |
1430 |
|
- Gene Symbol
- Lrrc8d
- Entrez Gene ID
- 231549
- Gene Name
- leucine rich repeat containing 8D
- Gene Aliases
- 2810473G09Rik, 4930525N13Rik, A930019F03, Lrrc5
- Chromosome Location
- Chr. 5: 105699969 - 105815215 on Build GRCm38
- UniGene ID
- Mm.23837
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK170452.1 |
- |
2 |
275 |
|
- Gene Symbol
- Sgip1
- Entrez Gene ID
- 73094
- Gene Name
- SH3-domain GRB2-like (endophilin) interacting protein 1
- Gene Aliases
- 3110007P09Rik, SGIP1alpha, gag
- Chromosome Location
- Chr. 4: 102759167 - 102972701 on Build GRCm38
- UniGene ID
- Mm.238094
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK043905.1 |
- |
|
2214 |
|
AK049616.1 |
- |
9 |
1215 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AK140518.1 |
- |
|
961 |
|
Chr. - - [-] |
View Full Product Details