Gene Transcripts
- Gene Symbol
- Chmp7
- Entrez Gene ID
- 105513
- Gene Name
- charged multivesicular body protein 7
- Gene Aliases
- 4930596K11Rik, 6330407G04Rik, AI450338, AW550775
- Chromosome Location
- Chr. 14: 69716979 - 69732570 on Build GRCm38
- UniGene ID
- Mm.159637
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK030270.1 |
- |
1 |
2169 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AV284316.2 |
- |
1 |
409 |
|
Chr. 14 - 69730203 - 69730875 [+] |
View Full Product Details