Gene Transcripts
- Gene Symbol
- 4930453O03Rik
- Entrez Gene ID
- 74885
- Gene Name
- RIKEN cDNA 4930453O03 gene
- Gene Aliases
- Chromosome Location
- Chr. 1: 88346937 - 88351722 on Build GRCm38
- UniGene ID
- Mm.449991
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK015456.1 |
- |
|
922 |
|
View Full Product Details