Gene Transcripts

Gene Symbol
Hottip
Entrez Gene ID
791364
Gene Name
Hoxa distal transcript antisense RNA
Gene Aliases
ENSMUSG00000055408, Gm15053, OTTMUSG00000018874
Chromosome Location
Chr. 6: 52262775 - 52267603 on Build GRCm38
UniGene ID
Mm.97466
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_110441.1 2 2317
NR_110442.1 2 1763
GenBank mRNA BC046496.1 - 2 1763

View Full Product Details