Gene Transcripts
- Gene Symbol
- Gm13630
- Entrez Gene ID
- 102639009
- Gene Name
- predicted gene 13630
- Gene Aliases
- OTTMUSG00000013101
- Chromosome Location
- Chr. 2: 66406076 - 66421589 on Build GRCm38
- UniGene ID
- Mm.403091
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK034270.1 |
- |
4 |
569 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB626272.1 |
- |
4 |
569 |
|
Chr. 2 - 66409495 - 66413899 [+] |
View Full Product Details