Gene Transcripts
- Gene Symbol
- A730082K24Rik
- Entrez Gene ID
- 101786
- Gene Name
- RIKEN cDNA A730082K24 gene
- Gene Aliases
- AU044236
- Chromosome Location
- Chr. 7: 114948650 - 114983967 on Build GRCm38
- UniGene ID
- Mm.395157
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_040317.1 |
|
4 |
1354 |
|
NR_040319.1 |
|
5 |
1332 |
|
GenBank mRNA |
AK043303.1 |
- |
2 |
902 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB258061.2 |
- |
1 |
317 |
|
Chr. 7 - 114983195 - 114983967 [+] |
GenBankmRNA |
DV661602.1 |
- |
1 |
315 |
|
Chr. 7 - 114983192 - 114983960 [+] |
View Full Product Details