Gene Transcripts
- Gene Symbol
- Nf1
- Entrez Gene ID
- 18015
- Gene Name
- neurofibromatosis 1
- Gene Aliases
- AW494271, E030030H24Rik, Nf-1
- Chromosome Location
- Chr. 11: 79335988 - 79581609 on Build GRCm38
- UniGene ID
- Mm.255596
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK053192.1 |
- |
1 |
3105 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AK162785.1 |
- |
|
1560 |
|
Chr. - - [-] |
View Full Product Details