Gene Transcripts
- Gene Symbol
- Spag9
- Entrez Gene ID
- 70834
- Gene Name
- sperm associated antigen 9
- Gene Aliases
- 3110018C07Rik, 4733401I23Rik, 4831406C20Rik, AW552012, JLP, JSAP2, JSAP2a, Jip4, Mapk8ip4, syd1
- Chromosome Location
- Chr. 11: 93996058 - 94126087 on Build GRCm38
- UniGene ID
- Mm.260737
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK079365.1 |
- |
1 |
420 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB130338.2 |
- |
1 |
193 |
|
Chr. 11 - 94044483 - 94045047 [+] |
View Full Product Details