Gene Transcripts
- Gene Symbol
- 4930471M09Rik
- Entrez Gene ID
- 75787
- Gene Name
- RIKEN cDNA 4930471M09 gene
- Gene Aliases
- 4930427C17Rik
- Chromosome Location
- Chr. 6: 91430460 - 91440854 on Build GRCm38
- UniGene ID
- Mm.30842
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_045980.1 |
|
3 |
768 |
|
GenBank mRNA |
AK015554.1 |
- |
3 |
769 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY715104.1 |
- |
3 |
769 |
|
Chr. 6 - 91430616 - 91440854 [-] |
View Full Product Details