Gene Transcripts
- Gene Symbol
- 4933440K10Rik
- Entrez Gene ID
- 71288
- Gene Name
- RIKEN cDNA 4933440K10 gene
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.275963
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK017135.1 |
- |
1 |
130 |
|
View Full Product Details