Gene Transcripts
- Gene Symbol
- Gm12002
- Entrez Gene ID
- 100503630
- Gene Name
- predicted gene 12002
- Gene Aliases
- OTTMUSG00000005167
- Chromosome Location
- Chr. 11: 12313365 - 12346088 on Build GRCm38
- UniGene ID
- Mm.442203
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK040873.1 |
- |
5 |
1462 |
|
View Full Product Details