Gene Transcripts
- Gene Symbol
- Gm12002
- Entrez Gene ID
- 100503630
- Gene Name
- predicted gene 12002
- Gene Aliases
- OTTMUSG00000005167
- Chromosome Location
- Chr. 11: 12313365 - 12346088 on Build GRCm38
- UniGene ID
- Mm.442203
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK040873.1 |
- |
3 |
326 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CK796166.1 |
- |
3 |
333 |
|
Chr. 11 - 12314532 - 12336060 [+] |
GenBankmRNA |
AI019723.1 |
- |
4 |
392 |
|
Chr. 11 - 12314533 - 12335830 [+] |
View Full Product Details