Gene Transcripts
- Gene Symbol
- 2810405F17Rik
- Entrez Gene ID
- 69975
- Gene Name
- RIKEN cDNA 2810405F17 gene
- Gene Aliases
- AI452280
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.31786
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK012993.1 |
- |
1 |
1041 |
|
AV136466.1 |
- |
1 |
206 |
|
View Full Product Details