Gene Transcripts
- Gene Symbol
- 4930466K18Rik
- Entrez Gene ID
- 74969
- Gene Name
- RIKEN cDNA 4930466K18 gene
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.442508
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK015515.1 |
- |
1 |
330 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BU610016.1 |
- |
1 |
414 |
|
Chr. 10 - 53614284 - 53614999 [+] |
View Full Product Details