Gene Transcripts
- Gene Symbol
- 1110064A23Rik
- Entrez Gene ID
- 68893
- Gene Name
- RIKEN cDNA 1110064A23 gene
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.69566
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK004365.1 |
- |
1 |
515 |
|
BC040515.1 |
- |
1 |
1578 |
|
BF180451.1 |
- |
1 |
607 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CF539586.1 |
- |
1 |
513 |
|
Chr. 2 - 31948549 - 31949081 [+] |
View Full Product Details