Gene Transcripts

Gene Symbol
5430434G16Rik
Entrez Gene ID
71385
Gene Name
RIKEN cDNA 5430434G16 gene
Gene Aliases
Chromosome Location
Chr. : - on Build
UniGene ID
Mm.440854
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK017390.1 - 1 509
AK139147.1 - 1 509
BE373635.1 - 1 493

View Full Product Details