Gene Transcripts
- Gene Symbol
- Gm4265
- Entrez Gene ID
- 100043150
- Gene Name
- predicted gene 4265
- Gene Aliases
- Chromosome Location
- Chr. 7: 129962165 - 129978669 on Build GRCm38
- UniGene ID
- Mm.334264
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_046077.1 |
|
4 |
629 |
|
GenBank mRNA |
AK137975.1 |
- |
4 |
630 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY723445.1 |
- |
4 |
626 |
|
Chr. 7 - 129962168 - 129978643 [+] |
View Full Product Details