Gene Transcripts
- Gene Symbol
- 5730412P04Rik
- Entrez Gene ID
- 70491
- Gene Name
- RIKEN cDNA 5730412P04 gene
- Gene Aliases
- Chromosome Location
- Chr. X: 136104603 - 136115420 on Build GRCm38
- UniGene ID
- Mm.396636
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_045424.1 |
|
3 |
450 |
|
GenBank mRNA |
AK017555.1 |
- |
3 |
452 |
|
View Full Product Details