Gene Transcripts
- Gene Symbol
- Galnt10
- Entrez Gene ID
- 171212
- Gene Name
- UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 10
- Gene Aliases
- AU018154, C330012K04Rik, GalNAc-T10, GalNAc-T9, Galnt9, ppGaNTase
- Chromosome Location
- Chr. 11: 57622407 - 57787501 on Build GRCm38
- UniGene ID
- Mm.271670
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK082774.1 |
- |
1 |
1219 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB399121.2 |
- |
1 |
637 |
|
Chr. 11 - 57726254 - 57726911 [+] |
View Full Product Details