Gene Transcripts

Gene Symbol
Gm16532
Entrez Gene ID
100042450
Gene Name
predicted gene, 16532
Gene Aliases
Chromosome Location
Chr. 7: 6415175 - 6431086 on Build GRCm38
UniGene ID
Mm.416919
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK029648.1 - 217


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BY714745.1 - 2 214 Chr. 9 - 21152538 - 21166880 [-]

View Full Product Details