Gene Transcripts
- Gene Symbol
- 4930517M08Rik
- Entrez Gene ID
- 75117
- Gene Name
- RIKEN cDNA 4930517M08 gene
- Gene Aliases
- Chromosome Location
- Chr. 17: 4634980 - 4637478 on Build GRCm38
- UniGene ID
- Mm.429393
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK015816.1 |
- |
3 |
615 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY715360.1 |
- |
3 |
615 |
|
Chr. 17 - 4634978 - 4637455 [+] |
View Full Product Details