Gene Transcripts
- Gene Symbol
- Zfand5
- Entrez Gene ID
- 22682
- Gene Name
- zinc finger, AN1-type domain 5
- Gene Aliases
- 2310057A04Rik, 5830475F03Rik, AA415485, Za20d2, Zfp216
- Chromosome Location
- Chr. 19: 21271279 - 21286840 on Build GRCm38
- UniGene ID
- Mm.292405
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK009947.1 |
- |
1 |
1384 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CO430396.1 |
- |
1 |
523 |
|
Chr. 19 - 21282976 - 21283709 [+] |
GenBankmRNA |
CX729734.1 |
- |
1 |
408 |
|
Chr. 19 - 21283090 - 21283699 [+] |
GenBankmRNA |
CN458386.1 |
- |
1 |
597 |
|
Chr. 19 - 21282901 - 21283643 [+] |
View Full Product Details