Gene Transcripts
- Gene Symbol
- Gm36017
- Entrez Gene ID
- 102639787
- Gene Name
- predicted gene, 36017
- Gene Aliases
- Chromosome Location
- Chr. 5: 44412732 - 44427652 on Build GRCm38
- UniGene ID
- Mm.108485
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK076918.1 |
- |
1 |
102 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB615740.1 |
- |
1 |
102 |
|
Chr. 5 - 44413664 - 44427590 [-] |
View Full Product Details