Gene Transcripts
- Gene Symbol
- 4930443G03Rik
- Entrez Gene ID
- 74873
- Gene Name
- RIKEN cDNA 4930443G03 gene
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.84387
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK015370.1 |
- |
1 |
589 |
|
View Full Product Details