Gene Transcripts
- Gene Symbol
- 3110035G12Rik
- Entrez Gene ID
- 73169
- Gene Name
- RIKEN cDNA 3110035G12 gene
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK014125.1 |
- |
1 |
1195 |
|
View Full Product Details