Gene Transcripts
- Gene Symbol
- Gm26744
- Entrez Gene ID
- 105246086
- Gene Name
- predicted gene, 26744
- Gene Aliases
- Chromosome Location
- Chr. 16: 21842111 - 21845850 on Build GRCm38
- UniGene ID
- Mm.438013
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK138627.1 |
- |
2 |
730 |
|
View Full Product Details