Gene Transcripts
- Gene Symbol
- LINC00632
- Entrez Gene ID
- 286411
- Gene Name
- long intergenic non-protein coding RNA 632
- Gene Aliases
- Chromosome Location
- Chr. X: 140709759 - 140772677 on Build GRCh38
- UniGene ID
- Hs.317627
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_028344.1 |
|
3 |
988 |
|
GenBank mRNA |
AK054978.1 |
- |
3 |
980 |
|
BQ343502.1 |
- |
1 |
258 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000370535.7 |
- |
3 |
980 |
|
Chr. X - 140709767 - 140714822 [+] |
View Full Product Details