Gene Transcripts
- Gene Symbol
- SH3BP5
- Entrez Gene ID
- 9467
- Gene Name
- SH3 domain binding protein 5
- Gene Aliases
- SAB, SH3BP-5
- Chromosome Location
- Chr. 3: 15254353 - 15341394 on Build GRCh38
- UniGene ID
- Hs.257761
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AA843106.1 |
- |
1 |
389 |
|
BX114836.1 |
- |
1 |
485 |
|
- Gene Symbol
- SH3BP5-AS1
- Entrez Gene ID
- 100505696
- Gene Name
- SH3BP5 antisense RNA 1
- Gene Aliases
- Chromosome Location
- Chr. 3: 15254184 - 15264498 on Build GRCh38
- UniGene ID
- Hs.745026
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_046084.1 |
|
1 |
1037 |
|
GenBank mRNA |
AL133111.1 |
- |
1 |
1037 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000420195.1 |
- |
1 |
1037 |
|
Chr. 3 - 15254184 - 15264493 [+] |
View Full Product Details