Gene Transcripts
- Gene Symbol
- LOC101927482
- Entrez Gene ID
- 101927482
- Gene Name
- uncharacterized LOC101927482
- Gene Aliases
- Chromosome Location
- Chr. 2: 196260024 - 196264204 on Build GRCh38
- UniGene ID
- Hs.191841
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_110225.1 |
|
3 |
882 |
|
NR_110226.1 |
|
3 |
871 |
|
GenBank mRNA |
BC018675.1 |
- |
2 |
761 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000433933.1 |
- |
2 |
871 |
|
Chr. 2 - 196260114 - 196264204 [+] |
View Full Product Details