Gene Transcripts
- Gene Symbol
- LOC101927143
- Entrez Gene ID
- 101927143
- Gene Name
- uncharacterized LOC101927143
- Gene Aliases
- Chromosome Location
- Chr. 1: 224208747 - 224213279 on Build GRCh38
- UniGene ID
- Hs.638720
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_110669.1 |
|
1 |
401 |
|
GenBank mRNA |
BC008027.1 |
- |
1 |
401 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000436706.1 |
- |
1 |
401 |
|
Chr. 1 - 224208747 - 224213279 [-] |
View Full Product Details