Gene Transcripts
- Gene Symbol
- LINC01440
- Entrez Gene ID
- 102723578
- Gene Name
- long intergenic non-protein coding RNA 1440
- Gene Aliases
- Chromosome Location
- Chr. 20: 55423043 - 55426791 on Build GRCh38
- UniGene ID
- Hs.446269
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000450623.6 |
- |
2 |
349 |
|
Chr. 20 - 55423011 - 55426790 [+] |
GenBankmRNA |
BC041864.1 |
- |
|
317 |
|
Chr. - - [-] |
View Full Product Details