Gene Transcripts
- Gene Symbol
- LINC00494
- Entrez Gene ID
- 284749
- Gene Name
- long intergenic non-protein coding RNA 494
- Gene Aliases
- Chromosome Location
- Chr. 20: 48359911 - 48370638 on Build GRCh38
- UniGene ID
- Hs.299080
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026958.1 |
|
5 |
1429 |
|
GenBank mRNA |
AL832259.1 |
- |
3 |
917 |
|
BC035164.2 |
- |
5 |
1392 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000416742.5 |
- |
5 |
1390 |
|
Chr. 20 - 48359950 - 48370629 [+] |
GENCODE |
ENST00000433094.1 |
- |
3 |
917 |
|
Chr. 20 - 48365647 - 48370629 [+] |
View Full Product Details