Gene Transcripts
- Gene Symbol
- LOC100507557
- Entrez Gene ID
- 100507557
- Gene Name
- uncharacterized LOC100507557
- Gene Aliases
- Chromosome Location
- Chr. 6: 145734869 - 145886585 on Build GRCh38
- UniGene ID
- Hs.124537
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000452617.5 |
- |
|
827 |
|
Chr. 6 - 145815241 - 145883439 [+] |
GENCODE |
ENST00000606388.5 |
- |
3 |
704 |
|
Chr. 6 - 145814912 - 145886585 [+] |
View Full Product Details