Gene Transcripts
- Gene Symbol
- LINC01419
- Entrez Gene ID
- 103352670
- Gene Name
- long intergenic non-protein coding RNA 1419
- Gene Aliases
- LVCAT7, TCONS_00014497
- Chromosome Location
- Chr. 8: 83403758 - 83408900 on Build GRCh38
- UniGene ID
- Hs.399852
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_122034.1 |
|
2 |
855 |
|
GenBank mRNA |
BC038578.1 |
- |
2 |
855 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000522365.1 |
- |
2 |
855 |
|
Chr. 8 - 83403758 - 83408900 [+] |
View Full Product Details