Gene Transcripts
- Gene Symbol
- LOC101927637
- Entrez Gene ID
- 101927637
- Gene Name
- uncharacterized LOC101927637
- Gene Aliases
- Chromosome Location
- Chr. 12: 127631269 - 127636467 on Build GRCh38
- UniGene ID
- Hs.170413
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_120451.1 |
|
4 |
554 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BX103747.1 |
- |
3 |
476 |
|
Chr. 12 - 127631309 - 127636106 [-] |
GENCODE |
ENST00000544251.5 |
- |
3 |
489 |
|
Chr. 12 - 127631248 - 127636119 [-] |
View Full Product Details