Gene Transcripts
- Gene Symbol
- LOC102723895
- Entrez Gene ID
- 102723895
- Gene Name
- uncharacterized LOC102723895
- Gene Aliases
- Chromosome Location
- Chr. 11: 104568489 - 104609302 on Build GRCh38
- UniGene ID
- Hs.48803
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_120585.1 |
|
4 |
411 |
|
GenBank mRNA |
DA109127.1 |
- |
4 |
411 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000545630.1 |
- |
4 |
411 |
|
Chr. 11 - 104568530 - 104609302 [-] |
View Full Product Details