Gene Transcripts
- Gene Symbol
- LOC101927292
- Entrez Gene ID
- 101927292
- Gene Name
- uncharacterized LOC101927292
- Gene Aliases
- Chromosome Location
- Chr. 12: 49911953 - 49926339 on Build GRCh38
- UniGene ID
- Hs.129345
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_110048.1 |
|
6 |
3138 |
|
GenBank mRNA |
BC034605.1 |
- |
6 |
3138 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000546821.5 |
- |
6 |
3138 |
|
Chr. 12 - 49911953 - 49926339 [+] |
View Full Product Details