Gene Transcripts
- Gene Symbol
- LOC101928414
- Entrez Gene ID
- 101928414
- Gene Name
- uncharacterized LOC101928414
- Gene Aliases
- Chromosome Location
- Chr. 15: 45251581 - 45279222 on Build GRCh38
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_120335.1 |
|
5 |
599 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000559003.5 |
- |
|
662 |
|
Chr. 15 - 45251686 - 45256002 [-] |
View Full Product Details