Gene Transcripts
- Gene Symbol
- LRRC37A8P
- Entrez Gene ID
- 100533789
- Gene Name
- leucine rich repeat containing 37 member A8, pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 17: 35904220 - 35911023 on Build GRCh38
- UniGene ID
- Hs.167595
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_126038.1 |
|
1 |
1281 |
|
GenBank mRNA |
AK128235.1 |
- |
1 |
1281 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000604907.1 |
- |
1 |
1281 |
|
Chr. 17 - 35893707 - 35911023 [-] |
View Full Product Details