Gene Transcripts
- Gene Symbol
- 4930465M20Rik
- Entrez Gene ID
- 74968
- Gene Name
- RIKEN cDNA 4930465M20 gene
- Gene Aliases
- 4930431H15Rik
- Chromosome Location
- Chr. 12: 107723743 - 107735488 on Build GRCm38
- UniGene ID
- Mm.359821
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_045973.1 |
|
3 |
791 |
|
GenBank mRNA |
AK015510.1 |
- |
3 |
791 |
|
View Full Product Details