Gene Transcripts
- Gene Symbol
- Cdrt4os1
- Entrez Gene ID
- 74884
- Gene Name
- CMT1A duplicated region transcript 4, opposite strand 1
- Gene Aliases
- 4930452L02Rik
- Chromosome Location
- Chr. 11: 62990453 - 63004134 on Build GRCm38
- UniGene ID
- Mm.363520
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK015448.1 |
- |
1 |
102 |
|
AK133035.1 |
- |
1 |
108 |
|
View Full Product Details