Gene Transcripts
- Gene Symbol
- Gm15679
- Entrez Gene ID
- 100873092
- Gene Name
- predicted gene 15679
- Gene Aliases
- OTTMUSG00000025218
- Chromosome Location
- Chr. 8: 99007127 - 99032740 on Build GRCm38
- UniGene ID
- Mm.414599
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_110579.1 |
|
3 |
1507 |
|
GenBank mRNA |
BC072565.1 |
- |
3 |
1507 |
|
View Full Product Details